Learn More
Grainger Wire Rope Oval Sleeve 1/16 in Copper
Supplier:  Grainger 16X774
Encompass_Preferred
The Loos & Co. Wire Rope Oval Sleeve is an oval-shaped wire rope end fitting made of drop forged steel with a zinc finish. It is designed to terminate wire rope by inserting the rope and compressing with a hydraulic or mechanical press. This oval sleeve model has a 16x7/8" size, fits 1/4" to 3/8" wire rope, and has a 1,400 lb working load limit. The fitting allows wire rope to be securely terminated in a loop for attachments.
Catalog No. 50-276-7027