Learn More
Grainger Wire Rope Oval Sleeve 1/2 in Copper
Supplier:  Grainger 16X786
Encompass_Preferred
The LOOS Wire Rope Oval Sleeve is an oval-shaped wire rope sleeve made of drop forged steel with a zinc finish. It is designed for securing the ends of 7x19 and 6x19 wire rope. This wire rope sleeve has an internal diameter of 1/2 inch, an external width of 27/32 inch, and an external length of 1-1/4 inch. The working load limit is 2,600 pounds. This wire rope end fitting allows wire rope to be looped and secured, providing a strong and reliable wire rope termination.
Catalog No. 50-276-8014