Learn More
Grainger Wire Rope Oval Sleeve 1/2 in Copper
Supplier:  Grainger 16X799
Encompass_Preferred
The LOOS Wire Rope Oval Sleeve is an oval-shaped wire rope end fitting made of drop forged steel with a zinc finish. It is designed to fit 1/4" wire rope and has an opening of 5/16" x 7/16". The working load limit is 1,260 lbs. This wire rope end fitting creates a loop and protects the end of the wire rope from unraveling. It can be used in a variety of wire rope applications to create an attachment point or loop. The oval shape allows for a smoother pull through pulleys compared to traditional round thimbles. This is a high quality wire rope fitting made in the USA by Loos & Co.
Catalog No. 50-276-6996