Learn More
Grainger Wire Rope Oval Sleeve 1/4 in Copper
Supplier:  Grainger 16X809
Encompass_Preferred
The LOOS Wire Rope Oval Sleeve is an oval-shaped wire rope sleeve made of galvanized steel. It has an inside diameter of 16mm and is designed for use with 6x7 and 6x19 wire rope. The sleeve is 80mm long and features a seamless construction with tapered ends for easy installation. This wire rope sleeve provides a secure connection and helps prevent unraveling of wire rope ends. It is suitable for a variety of applications including industrial rigging, marine environments, overhead lifting, and more. The oval shape allows for a lower profile connection.
Catalog No. 50-276-7791