Learn More
Grainger Wire Rope Oval Sleeve 1/4 in Copper
Supplier:  Grainger 16X794
Encompass_Preferred
The Locoloc Wire Rope Oval Sleeve is an oval-shaped wire rope sleeve made of zinc-plated steel. It is designed to fit 1/4" wire rope and has an opening width of 0.27". The working load limit is 1,260 lbs. This wire rope sleeve can be used for various rigging and hoisting applications to protect the end of wire rope from fraying. The oval shape provides a smooth profile. Overall, this is a high-quality wire rope end sleeve for 1/4" diameter wire rope with a 1,260 lb working load limit.
Catalog No. 50-276-6992