Learn More
Grainger Wire Rope Oval Sleeve 1/4 in Copper
Supplier:  Grainger 16X808
Encompass_Preferred
The LOOS Wire Rope Oval Sleeve is an oval-shaped wire rope sleeve made of galvanized steel. It has an inside diameter of 1/2", is 1-1/4" long, and weighs 0.04 lbs. This wire rope sleeve is designed to fit 1/2" diameter 7x7 or 7x19 wire rope and can be used for a variety of wire rope repair applications. The galvanized coating provides corrosion resistance. This wire rope sleeve meets military specification MIL-S-781. It is sold individually.
Catalog No. 50-276-7919