Learn More
Grainger Wire Rope Oval Sleeve 1/4 in Copper
Supplier:  Grainger 16X782
Encompass_Preferred
The LOOS Wire Rope Oval Sleeve is an oval-shaped wire rope sleeve made of drop forged steel. It has a working load limit of 9,800 lbs and is designed for 5/16" 6x19 IWRC right regular lay galvanized wire rope. The sleeve's length is 1-3/4" and width is 15/16". It can be used for various rigging and hoisting applications. The sleeve is zinc plated for corrosion resistance. It meets military specification MIL-S-781 and features the Crosby brand, known for quality rigging and lifting hardware. The sleeve provides a secure end connection and protects the wire rope from fraying.
Catalog No. 50-276-7026