Learn More
Grainger Wire Rope Oval Sleeve 1/4 in Copper
Supplier:  Grainger 16X795
Encompass_Preferred
The LOOS Wire Rope Oval Sleeve is an oval-shaped wire rope sleeve made of galvanized steel. It has an inside diameter of 1/2", fits 1/8" wire rope, and is 7/8" long. This wire rope sleeve is designed to protect the ends of wire rope from fraying and unraveling. It can be used in a variety of wire rope termination applications. The galvanized coating provides corrosion resistance. This wire rope sleeve meets military specification MIL-S-781. It is sold individually.
Catalog No. 50-276-6866