Learn More
Grainger Wire Rope Oval Sleeve 1/8 in Copper
Supplier:  Grainger 16X804
Encompass_Preferred
The LOOS Wire Rope Oval Sleeve is an oval-shaped wire rope sleeve made of drop forged steel. It has a working load limit of 9,800 lbs and is designed for 5/16" 6x19 or 6x37 IPS IWRC right regular lay galvanized wire rope. The sleeve's length is 1-3/4", width is 1-1/8", thickness is 17/32", and weight is 0.19 lbs. This wire rope end fitting provides a spliced eye termination and is used for rigging and hoisting applications. The sleeve is zinc plated for corrosion resistance. It should be installed using the proper swaging technique for safe and secure wire rope connections.
Catalog No. 50-276-6847