Learn More
Grainger Wire Rope Oval Sleeve 1/8 in Copper
Supplier:  Grainger 16X805
Encompass_Preferred
The Grainger 1/8" Copper Wire Rope Oval Sleeve is made of copper and designed to fit 1/8" wire rope. It measures 1-1/4" long x 5/8" wide and has an oval cross section to grip the rope. The copper construction provides durability and conductivity. This wire rope sleeve can be used for electrical connections and helps protect against corrosion and weathering. It is rated up to 600 volts and is commonly used for grounding and bonding applications. The oval design allows for secure installation without special tools. This is a high quality copper wire rope sleeve for 1/8" wire rope.
Catalog No. 50-276-6868