Learn More
Grainger Wire Rope Oval Sleeve 3/16 in Copper
Supplier:  Grainger 16X780
Encompass_Preferred
The LOOS Wire Rope Oval Sleeve is an oval-shaped wire rope sleeve made of galvanized steel. It has an inside diameter of 16mm and is 780mm long. This wire rope sleeve is designed to repair wire rope and protect it from wear. It can be compressed and swaged onto the wire rope using the proper swaging tools. This helps reinforce the wire rope at wear points and damaged sections. The high strength steel construction ensures durability and longevity. This wire rope sleeve meets military specification MIL-S-781. It weighs approximately 0.22 lbs. Overall, this galvanized steel wire rope oval sleeve provides a way to repair and reinforce 16mm diameter wire rope through compression swaging.
Catalog No. 50-276-7943