Learn More
Grainger Wire Rope Oval Sleeve 3/16 in Copper
Supplier:  Grainger 16X806
Encompass_Preferred
The LOOS Wire Rope Oval Sleeve is an oval-shaped wire rope sleeve made of drop forged steel with a zinc plating. It is designed to fit 1/4" wire rope and has an opening width of 0.27". The working load limit is 1,260 lbs. This wire rope sleeve can be used for a variety of securing and rigging applications to protect the end of wire rope from fraying. The oval shape provides a smooth, finished look. Overall, this is a high-strength, corrosion-resistant wire rope end fitting for 1/4" diameter wire rope.
Catalog No. 50-276-7920