Learn More
Grainger Wire Rope Oval Sleeve 7/16 in Copper
Supplier:  Grainger 16X798
Encompass_Preferred
The Grainger 7/16" Copper Oval Sleeve is a wire rope fitting designed to join two wire rope ends together. It is made from copper with a bright finish and features an oval shape with two holes to accept 7/16" wire rope. This wire rope sleeve measures 1-1/4" long x 1-1/8" wide x 15/16" high. It is rated up to 8,600 lb breaking strength. The copper construction provides good conductivity and corrosion resistance. This wire rope fitting can be used for a wide variety of wire rope end connections in industrial, commercial and marine applications.
Catalog No. 50-276-6871