Learn More
Grainger Wire Rope Stop Sleeve 1/16 in Copper
Supplier:  Grainger 16X816
Encompass_Preferred
The Loos Wire Rope Stop Sleeve Stop is a wire rope fitting designed to prevent the end of a wire rope from unraveling. It is made from drop-forged carbon steel and zinc plated for corrosion resistance. This wire rope stop has a working load limit of 1,250 lbs and is designed for 1/4" 7x7 or 7x19 wire rope. It meets military specification MIL-S-781. Overall length is 1-1/4" and the barrel length is 13/16". This wire rope stop provides a permanent stop and fits into tight areas. It is for use on stationary wire rope ends.
Catalog No. 50-276-6802