Learn More
Grainger Wire Rope Stop Sleeve 1/16 in Copper
Supplier:  Grainger 16X814
Encompass_Preferred
The Loos Wire Rope Stop Sleeve Stop is a wire rope termination fitting designed to prevent fraying and unraveling of wire rope ends. It is made from drop forged steel and zinc plated for corrosion resistance. This stop sleeve can be used with 6x7 and 6x19 wire rope constructions from 1/8" to 1/2" diameter. It installs by sliding over the bitter end of the wire rope and compressing with a swaging tool to permanently secure it in place. This stop sleeve provides a finished look and helps prevent accidents due to frayed wire rope ends.
Catalog No. 50-276-7972