Learn More
Grainger Wire Rope Stop Sleeve 1/4 in Copper
Supplier:  Grainger 16X821
Encompass_Preferred
The Loos Wire Rope Stop Sleeve Stop is a wire rope termination device designed to prevent fraying of wire rope ends. It is made from drop-forged steel and zinc plated for corrosion resistance. This wire rope stop can be used on 6x7 and 6x19 wire rope constructions from 1/8" to 1/2" diameter. It installs by sliding over the bitter end of the wire rope and compressing with a hammer to grip the rope. This wire rope stop provides a finished look to wire rope ends.
Catalog No. 50-276-6994