Learn More
Grainger Wire Rope Stop Sleeve 3/8 in Copper
Supplier:  Grainger 16X824
Encompass_Preferred
The Grainger Approved Wire Rope Stop Sleeve is designed to prevent fraying and unraveling of wire rope ends. It is constructed of drop-forged steel with a zinc finish for corrosion resistance. This wire rope stop sleeve accommodates 1/4" to 1/2" diameter 7x7 and 7x19 wire rope and features two set screws to securely grip the wire rope. Overall length is 2-1/2" and weight is 0.8 lbs. This wire rope stop sleeve provides an economical way to help protect against accidents caused by frayed wire rope ends in industrial settings.
Catalog No. 50-276-6995