Learn More
Grainger Wire Rope Stud 5/16 -24 LH Thread 3/16in
Supplier:  Grainger 16X697
Encompass_Preferred
The Grainger 3/16" Swage Stud Fittings for Wire Rope are high-strength fittings used to terminate wire rope for lifting and rigging applications. They feature a forged carbon steel construction with zinc plating for corrosion resistance. The stud shank diameter is 3/16" with UNC thread. Load ratings vary by stud size from 1,740 lbs to 9,240 lbs. They meet ASME B30.26 specifications and are manufactured in the USA. Swage studs provide a secure wire rope termination and allow attachment of various rigging components like hooks, shackles and eye bolts.
Catalog No. 50-276-5700