Learn More
Grainger Wire Rope Stud 7/16 -20 RH Thread 9/32in
Supplier:  Grainger 16X703
Encompass_Preferred
The Grainger 7/16"-20 swage stud fittings for wire rope are high-strength forged steel fittings used to terminate wire rope. They feature a swaged collar design that grips the wire rope for a strong and secure connection. The fittings have a zinc plating for corrosion resistance and rust prevention. They are available in a variety of sizes including 7/16"-20 thread to accommodate different wire rope diameters. The working load limit is 2,600 lbs providing reliable performance under load. Overall, these swage stud fittings provide a durable and dependable wire rope termination solution.
Catalog No. 50-276-7942