Learn More
Grainger Wire Rope Thimble 1/8 in Rope Dia Steel
Supplier:  Grainger 16X761
Encompass_Preferred
The Loos & Co. Wire Rope Thimble is made of galvanized steel with a 3/32 inch inside diameter. It is designed to fit 3/32 inch wire rope and protect it from wear. The thimble has a closed eye and is used to form eyes on wire rope for use with shackles, hooks, and other hardware. It provides a hard wearing surface to prevent crushing and deformation of the wire rope. This wire rope thimble is galvanized for corrosion resistance. It meets military specification MIL-T-781J and is made in the USA by Loos & Co., a leading manufacturer of wire rope fittings.
Catalog No. 50-276-7086