Learn More
Grainger Wire Rope Thimble 3/16in Rope Dia Steel
Supplier:  Grainger 16X763
Encompass_Preferred
The Loos Wire Rope Thimble is a galvanized steel thimble with a 3/8" inside diameter that is compatible with 3/8" wire rope. It has a working load limit of 1,260 lbs and meets military specification MIL-T-781J. The thimble has a closed eye and is used to prevent fraying of wire rope. It can be used in rigging and sling applications. The thimble is made in the USA and sold individually.
Catalog No. 50-276-7020