Learn More
Grainger Wire Rope Thimble 3/8 in Rope Dia SS
Supplier:  Grainger 16X773
Encompass_Preferred
The Loos Wire Rope Thimble is a 3/8 inch galvanized steel thimble with a standard barrel design. It is compatible with 3/8 inch wire rope and has a working load limit of 1,260 lbs. The thimble has a length of 1-1/4 inches, width of 13/16 inches, barrel diameter of 1/2 inch, and weighs 0.06 lbs. It is made in the USA and conforms to military specification MIL-T-781J. The thimble helps prevent fraying of wire rope ends for safe, long-lasting use.
Catalog No. 50-276-5120