Learn More
Grainger Wire Rope Thimble 5/16 in Rope Dia SS
Supplier:  Grainger 16X772
Encompass_Preferred
The Loos Wire Rope Thimble is a galvanized steel thimble with a 9/32" inside diameter. It is designed to fit wire rope and help prevent fraying. The thimble has a closed eye and is made in the USA. It can be used in rigging and sling applications to protect wire rope from bending stresses. The thimble is rated up to 5 tons working load limit. It meets military specification MIL-T-81490 and features the Loos & Co. logo stamped on the side. Overall, this thimble provides reinforcement and protection for wire rope ends.
Catalog No. 50-276-8043