Learn More
ATCC WISH; HeLa Contaminant; Human (Homo Sapiens)
Supplier:  ATCC CCL25
Encompass
The WISH human cell line contains HeLa marker chromosomes and were derived via HeLa contamination. This cell line was deposited by L Hayflick.
Catalog No. 50-238-4442