Learn More
Cayman Chemical WISKOSTATIN 10MG
SDP
Supplier:  Cayman Chemical 1504710 MG
Encompass_Preferred
A cell-permeable, selective inhibitor of N-WASP (IC50 = ~10 μM); directly associates with the GTPase-binding domain of N-WASP, preventing its activation by Cdc42 and PIP2; interferes with membrane transport, endocytosis, and neurological development
Catalog No. 50-297-2253