Learn More
TARGETMOL CHEMICALS INC WISKOSTATIN 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T216410MG
Encompass_Preferred
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Wiskostatin is a selective inhibitor of N-WASP a ubiquitously expressed member of the Wiskott-Aldrich Syndrome protein (WASp) family that regulates actin polymerization. purity: 99%
Catalog No. 50-257-7209