Learn More
World Precision Instrument AutoLCI Adapter for 35mm FluoroDish, Dual
SDP
Supplier:  World Precision Instrument 505628
Encompass
With EVOM™ AutoLCI, cells cultured in different vessel types can be imaged. Interchangeable vessel adapters can accommodate various vessel types including well-plate, culture flask, and culture dish.  The system includes a well-plate adapter, but you can buy separately different adapters for a variety of vessels used.
Catalog No. 50-252-1270