Learn More
Apexbio Technology LLC Withaferin A 5119-48-2 1mg
SDP
Supplier:  Apexbio Technology LLC B71991
Encompass_Preferred
Withaferin A (CAS 5119-48-2) is a steroidal lactone isolated from the plant Withania somnifera This compound inhibits the activation of nuclear factor kappa B (NF- B) and interacts with vimentin a cytoskeletal protein leading to modulation of inflammatory and oncogenic signaling pathways Experimental studies have shown that Withaferin A suppresses the shedding of endothelial protein C receptor (EPCR) indicating an additional regulatory role in vascular biology These properties support its use as a research tool for investigating the molecular mechanisms underlying tumor progression and inflammatory diseases
Catalog No. 50-000-87858