Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Withangulatin A | 120824-03-5 | 526.62 | 1 MG
SDP

Supplier:  Medchemexpress LLC HYN103031MG

Encompass_Preferred

Withangulatin A is an inhibitor for COX-2. It also inhibits the MAPK, NF-κB, Akt/mTOR/p70S6K pathway. It exhibits antitumor, anti-inflammatory, and trypanocidal activities.

  • Inhibits COX-2.
  • Inhibits MAPK, NF-κB, Akt/mTOR/p70S6K pathway.
  • Exhibits antitumor activity.
  • Exhibits anti-inflammatory activity.
  • Exhibits trypanocidal activity.
  • Purity: 99.76%.
  • Appearance: solid, white to off-white.

Catalog No. 50-004-16920


May include imposed supplier surcharges.
Only null left
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.