Learn More
Medchemexpress LLC Withangulatin A | 120824-03-5 | 526.62 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYN103035MG
Encompass_Preferred
Withangulatin A is an inhibitor for COX-2 that demonstrates antitumor, anti-inflammatory, and trypanocidal activities. It inhibits MAPK, NF-κB, Akt/mTOR/p70S6K pathway, and various cellular processes in vitro and in vivo.
  • Inhibits proliferation of mouse T lymphocytes with an EC50 of 2.89 μM
  • Reduces LPS-induced secretion of IL-2, IFN-γ, and IL-6
  • Suppresses expression of COX-2 and PGE2 in mouse T lymphocytes
  • Arrests cell cycle at S phase and induces apoptosis in LX-2 cells
  • Inhibits TGF-β-stimulated collagen I gene expression and phosphorylation of Smad2/3
  • Binds to peroxiredoxin 6 (PRDX6) and inhibits GPx and iPLA2 activities
  • Promotes generation of ROS
  • Exhibits anti-inflammatory activity in mouse xylene-induced ear swelling models
  • Shows antitumor effect in mouse H1975 xenograft models
Catalog No. 50-004-16922