Learn More
Medchemexpress LLC WIZ degrader 4 | 2879250-22-1 | 99.4% | 371.43 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY16382010MM/1ML
Encompass_Preferred
WIZ degrader 4 is a WIZ degrader with an AC50 value of 160 nM. It is suitable for investigating sickle cell disease (SCD) and thalassemia.
  • AC50 value of 160 nM
  • Investigates sickle cell disease (SCD)
  • Investigates thalassemia
  • Purity of 99.37%
Catalog No. 50-003-90732