Learn More
Research Products International Corp Wizard Mini-Fuge, Quiet and Vibration Free
SDP
Supplier:  Research Products International Corp 400196
Encompass
Personal WIZARD mini-Fuge performs quick spins up to 6,000 rpm at the workbench, in a cold room, or fume hood. Supplied with two convenient snap on rotors that hold six 1.5-2.0 tubes or two strips of 8 x 0.2 ml tubes. Also accommodates skirted tubes and micro-filtration tube assemblies. Designed for years of rugged and dependable performances.
Compact, space saving
Spins 0.2, 1.5, or 2.0 ml micro-tubes
Quiet and vibration free
Catalog No. 50-197-7287