Learn More
Medchemexpress LLC WJ-39 | 3009908-95-3 | 99.6% | 430.32 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY16232510MM/1ML
Encompass_Preferred
WJ-39 is an orally active aldose reductase (AR) inhibitor that improves tubular damage in diabetic nephropathy rats. It functions by activating PINK1/Parkin signaling, promoting mitophagy, and attenuating apoptosis.
  • Orally active aldose reductase inhibitor
  • Ameliorates renal tubular injury in diabetic nephropathy
  • Activates PINK1/Parkin signaling
  • Promotes mitophagy
  • Attenuates apoptosis
  • Relevant to inflammation and metabolic diseases
  • Associated with apoptosis, autophagy, and mitochondrial metabolism
Catalog No. 50-003-92979