Learn More
Medchemexpress LLC WJ-39 | 3009908-95-3 | >99.6% | 430.32 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY162325100MG
Encompass_Preferred
WJ-39 is an orally active aldose reductase (AR) inhibitor. It improves tubular damage in diabetic nephropathy rats by activating PINK1/Parkin signaling, promoting mitophagy, and attenuating apoptosis.
  • Orally active aldose reductase (AR) inhibitor.
  • Ameliorates renal tubular injury in diabetic nephropathy.
  • Activates PINK1/Parkin signaling.
  • Promotes mitophagy.
  • Attenuates apoptosis.
Catalog No. 50-003-92289