Learn More
Medchemexpress LLC Wl47 (TFA) | 99.7% | 1942.32 | 5 MG
SDP
Supplier:  Medchemexpress LLC 5MGHYP2288A
Encompass_Preferred
WL47 TFA is a high-affinity cavolin-1 (CAV1) ligand that potently disrupts CAV1 oligomers. It shows selectivity for CAV1 over BSA, casein, and HEWL, and is used for studying caveolin-1 function. It is also 80% smaller in length than the original T20 parent sequence.
  • High-affinity cavolin-1 (CAV1) ligand (Kd=23 nM)
  • Potently disrupts CAV1 oligomers
  • Exhibits selectivity for CAV1 over BSA, casein, and HEWL
  • Used for studying caveolin-1 function
  • 80% smaller in length than original T20 parent sequence
Catalog No. 50-003-93448