Learn More
Selleck Chemical LLC WM-1119 5mg 2055397-28-7
SDP
Supplier:  Selleck Chemical LLC S87765MG
Encompass_Preferred
WM-1119 is a highly potent, selective inhibitor of lysine acetyltransferase KAT6A with IC50 of 0.25 ?M. It is 1,100-fold and 250-fold more active against KAT6A than against KAT5 or KAT7, respectively. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8264