Learn More
Apexbio Technology LLC WM-2474 5mg
SDP
Supplier:  Apexbio Technology LLC A87805
Encompass_Preferred
WM-2474 is a negative control compound structurally related to the histone acetyltransferase (HAT) inhibitor WM-8014 with minimal activity against the KAT6A/B enzymes It exhibits negligible inhibitory activity with a reported IC value exceeding 125 M WM-2474 does not alter cellular proliferation underscoring its lack of significant impact on KAT6A/B-mediated processes Based on these pharmacological properties WM-2474 is used as a control in research involving epigenetic regulation and tumor cell biology particularly to assess non-specific effects linked to the compound s structural features rather than active HAT inhibition
Catalog No. 50-224-0207