Learn More
Medchemexpress LLC WM-586 | 2894832-05-2 | 98.7% | 467.46 | C20H20F3N5O3S | 10 MG
SDP
Supplier:  Medchemexpress LLC HY15372810MG
Encompass_Preferred
WM-586 is a covalent small-molecule inhibitor of WDR5 used as a research probe to disrupt WDR5-MYC interactions and study epigenetic regulation in cancer models. It is supplied as a characterized, high-purity compound for in vitro and preclinical research.
  • Covalent WDR5 inhibitor targeting the WBM binding pocket.
  • Disrupts WDR5-MYC interaction for epigenetic research.
  • Reported IC50 approximately 101 nM in biochemical assays.
  • High chemical purity and enantiomeric excess.
  • Available in multiple milligram-scale pack sizes for research use.
  • Storage: 4°C under inert atmosphere; in solvent: -80°C (6 months) or -20°C (1 month).
  • Comprehensive analytical data provided, including COA, HNMR, LCMS, and HPLC.
Catalog No. 50-002-87047