Learn More
Medchemexpress LLC WM-662 | 1308257-47-7 | C19H18ClN5O4 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY15484710MM/1ML
Encompass_Preferred
WM-662 is an inhibitor of the WDR5-MYC interaction, with an IC50 of 18 μM. It shows potential for research in various disease areas including cancer, aging, and neurodegenerative disorders, and is intended for research use only. This compound is part of the epigenetics research focusing on WDR5.
  • Inhibits WDR5-MYC interaction with an IC50 of 18 μM
  • Potential for research in cancer
  • Potential for research in aging
  • Potential for research in neurodegenerative disorders
  • For research use only
Catalog No. 50-003-76058