Learn More
Medchemexpress LLC WM-662 | 1308257-47-7 | C19H18ClN5O4 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY154847
Encompass_Preferred
WM-662 is an inhibitor of the WDR5-MYC interaction, with an IC50 of 18 μM. It shows potential for research in cancer, aging, and neurodegenerative disorders.
  • WDR5-MYC interaction inhibitor
  • Potential for research in cancer, aging, and neurodegenerative disorders
Catalog No. 50-003-75035