Learn More
Selleck Chemical LLC WM-8014 2mg
SDP
Supplier:  Selleck Chemical LLC S66552MG
Encompass_Preferred
WM-8014 is a highly potent inhibitor of histone acetyltransferase KAT6A with an IC50 of 8 nM. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-217-3198