Learn More
Medchemexpress LLC WNK-IN-11-d3 | 2123483-49-6 | 99.8% | 465.41 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY112094S10MG
Encompass_Preferred
WNK-IN-11-d3 is an orally active, selective, and potent With-No-Lysine (WNK) kinase inhibitor. It is effective at regulating cardiovascular homeostasis. This compound can be used as a tracer or an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Orally active, selective, and potent WNK kinase inhibitor
  • Regulates cardiovascular homeostasis
  • Can be used as a tracer
  • Functions as an internal standard for quantitative analysis (NMR, GC-MS, LC-MS)
  • Improves rat PK profile, including lower clearance and enhanced oral bioavailability
  • Significantly reduces systolic blood pressure in mice
  • Induces dose-dependent diuresis, natriuresis, and kaliuresis
  • Reduces blood pressure, stroke volume, and total peripheral resistance; increases heart rate
  • Efficacious in rodent models of hypertension and volume overload
Catalog No. 50-004-37137