Learn More
Eurofins Discoverx WNK2, active, 250 µg. (14-678M)
Supplier:  Eurofins Discoverx 14678M
Encompass
N-terminal 6xHis-tagged recombinant, human WNK2 amino acids 166-489. For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-199-9174