Learn More
Selleck Chemical LLC WNK463 5mg 2012607-27-9
SDP
Supplier:  Selleck Chemical LLC S83585MG
Encompass_Preferred
WNK463 is a pan-WNK-kinase inhibitor. It potently inhibits the in vitro kinase activity of all four WNK family members (WNK1, WNK2, WNK3, and WNK4). *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-5980