Learn More
Medchemexpress LLC Wnt pathway inhibitor 3 | 663213-98-7 | 99.0% | 457.2800 g/mol | C21H17BrN2O5 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY15375010MG
Encompass_Preferred
Wnt pathway inhibitor 3 is a small-molecule research reagent that inhibits the Wnt signaling pathway with an IC50 of 45 nM and displays antiproliferative activity in cellular assays. It is supplied as a high-purity solid for laboratory research (CAS 663213-98-7; formula C21H17BrN2O5; MW 457.28 g/mol).
  • Potent Wnt pathway inhibition with IC50 of 45 nM.
  • Demonstrates antiproliferative activity in cellular assays.
  • High purity (98.96%) suitable for research use.
  • Supplied as a solid in small research quantities (5 mg-100 mg).
  • Chemical formula C21H17BrN2O5 and molecular weight 457.28 g/mol.
Catalog No. 50-002-87007