Learn More
Research Products International Corp Wobble-Not SERO Pipets, 5ml, 200/pk
SDP
Supplier:  Research Products International Corp 163250
Encompass
Wobble-Not Serological Pipets feature innovative plug-end design that require less force when inserting to pipet controller. Pipets offer an excellent secure seal, maximum stability, better accuracy and ultra-clear markings. Supplied individually wrapped (paper/plastic) with porous polypropylene filter and ±2% accuracy at full volume.
Snug, tight fit
Drip free
RNase/DNase free
Sterile, non-pyrogenic
Catalog No. 50-222-9458