Learn More
Enzo Life Sciences Wogonin (25mg). CAS: 632-85-9
SDP
Supplier:  Enzo Life Sciences ALX385033M025
Encompass_Preferred
Cell permeable and orally available flavonoid. Induces apoptosis in tumor cells but not in healthy cells by increasing peroxide production. Displays anti-inflammatory properties. Suppresses the release of nitric oxide (NO) by inducible nitric oxide synthase (iNOS; NOS II), PGE2 by cyclooxygenase-2 (COX-2), proinflammatory cytokines, MCP-1 gene expression and NF-κB activation. Antioxidant. Alternative name: 5,7-Dihydroxy-8-methoxyflavone. Purity: ≥98% (HPLC). Appearance: White to yellow solid. Solubility: Soluble in DMSO (10mg/ml). Long Term Storage: +4°C.
Catalog No. 50-200-9060