Learn More
AdipoGen Woollins Reagent
SDP
Supplier:  AdipoGen CDXW0019G005
Encompass_Preferred
Chemical. CAS 122039-27-4. Formula C12H10P2Se4. MW BD9837. Woollins Reagent is an organophosphorus compound that finds widespread utilization as a reagent in organic synthesis. Woollins Reagent participates in diverse reactions, such as yielding spiro heterocycles when combined with benzoic hydrazides, generating secondary amines, forming 10-membered phosphorus-selenium heterocycles in conjunction with aromatic diols, and facilitating ring-opening deselenation and phosphorus-phosphorus coupling. Woollins? Reagent has contributed to the synthesis of biologically active compounds, including anticancer drugs, antibiotics and antiviral agents.
Catalog No. 50-255-9906