Learn More
Grainger COTTERMAN Rolling Work Platform, 3 Steps
Supplier:  Grainger 20Z542
Encompass_Preferred
The Cotterman Rolling Work Platform is a 3-step steel rolling work platform with a weight capacity of 300 lbs. It features a 21.5" x 19.5" top work surface and has 3 large 8" rubber casters for easy mobility. The total height is 32.5" and each step is 8" deep. The steps are open for easy access. This provides the user a sturdy and mobile work surface for tasks that require extra height reach.
Catalog No. NC2739908