Learn More
Grainger Workbench Laminate 60 W 24 D
Supplier:  Grainger 16Y073
Encompass_Preferred
The Pro-Line L-Shaped Workbench with Adjustable Height is an industrial-grade workbench made from heavy-gauge steel with a powder-coated finish for durability. It features an L-shaped design with two work surfaces, providing ample workspace. The workbench has adjustable-height legs, allowing you to customize the height from 30" to 42" high. Other details include built-in lower shelves for storage, pre-drilled holes for adding accessories like bins and hooks, and leveling feet. This heavy-duty workbench can support up to 2500 lbs and is ideal for garage, shop, or industrial use.
Catalog No. 50-276-5555